Skip to Content
Merck
All Photos(2)

Key Documents

A8056

Sigma-Aldrich

3-Amino-1,2,4-triazole

powder, ≥95% (TLC)

Synonym(s):

1,2,4-Triazol-3-amine, 3-AT, Amitrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
Beilstein:
107687
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3-Amino-1,2,4-triazole, ≥95% (TLC)

biological source

synthetic (organic)

Quality Level

Assay

≥95% (TLC)

form

powder

potency

30 μM Ki

mp

150-153 °C (lit.)

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Nc1nc[nH]n1

InChI

1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)

InChI key

KLSJWNVTNUYHDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Amino-1,2,4-triazole is a herbicide effective against weeds especially bermuda and quack grass. It enters plant circulation through roots and is majorly present in the meristem. 3-Amino-1,2,4-triazole inhibits chlorophyll synthesis resulting in chlorotic leaves and albino plants. In addition, it also affects the growth of the plants. It is exploited to destroy unwanted weeds. It is synthesized from aminoguanidine formate and corresponds to a molecular weight of 84 Da.

Application

3-Amino-1,2,4-triazole has been used:
  • as an organelle-specific perturbant in human embryonic kidney (HEK293T) cells
  • as histidine biosynthesis inhibitor in medium for screening co-transformants from yeast two-hybrid interactions
  • for preservation of midgut sample from honey bee
  • for the determination of tryptophan in proteins.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Understanding organellar protein folding capacities and assessing their pharmacological modulation by small molecules
Sharma R, et al.
European Journal of Cell Biology, 97(2), 114-125 (2018)
Thermoanalytical study of the pesticide 3-amino-1, 2, 4-triazole
Sanchez-Soto P, et al.
Journal of Thermal Analysis and Calorimetry, 45(5), 1189-1197 (1995)
Nt-RhoGDI2 regulates Rac/Rop signaling and polar cell growth in tobacco pollen tubes
Klahre U, et al.
The Plant Journal, 46(6), 1018-1031 (2006)
The Biology and Control of Weeds in Sugarcane, 88-88 (2012)
Aarthi Putarjunan et al.
Nature plants, 5(7), 742-754 (2019-06-27)
Cell fate in eukaryotes is controlled by mitogen-activated protein kinases (MAPKs) that translate external cues into cellular responses. In plants, two MAPKs-MPK3 and MPK6-regulate diverse processes of development, environmental response and immunity. However, the mechanism that bridges these shared signalling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service