Skip to Content
Merck
All Photos(2)

Key Documents

207896

Sigma-Aldrich

Copper(II) carbonate basic

reagent grade

Synonym(s):

Cupric carbonate basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCO3 · Cu(OH)2
CAS Number:
Molecular Weight:
221.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

reagent grade

Quality Level

form

powder

concentration

50.0-57.0% Cu (EDTA titration)

SMILES string

[Cu++].O[Cu]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

ZMMDPCMYTCRWFF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) carbonate basic is naturally found in azurite and malachite. On heating, it undergoes decomposition to form copper(II) oxide, carbon dioxide and water. Due to its high initial discharge capacity, copper(II) carbonate (basic) is being considered as a promising candidate as anode materials for lithium-ion batteries.

Application

Copper(II) carbonate basic (Cupric carbonate basic, Copper(II) basic carbonate) may be used in the preparation of colloidal copper.
Copper(II) carbonate basic along with different ligands form an efficient catalytic system for ′on water′ β-borylation of Michael acceptors, alkenes and alkynes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basic CuCO3/ligand as a new catalyst for'on water'borylation of Michael acceptors, alkenes and alkynes: application to the efficient asymmetric synthesis of ?-alcohol type sitagliptin side chain.
Stavber G and Casar Z.
Applied Organometallic Chemistry, 27(3), 159-165 (2013)
Preparation and characterization of basic carbonates as novel anode materials for lithium-ion batteries.
Shao L, et al.
Ceramics International, 40(2), 3105-3116 (2014)
Gerrans GC, et al.
Sasol Encyclopaedia of Science and Technology (2004)
Daintith J
The facts on file dictionary of chemistry, 60-60 (2014)
Preparation and characterization of uniform particles of pure and coated metallic copper.
Hsu WP, et al.
Powder Technology, 63(3), 265-275 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service