Skip to Content
Merck
All Photos(2)

Key Documents

379956

Sigma-Aldrich

Cobalt(II) carbonate hydrate

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCO3·xH2O
CAS Number:
Molecular Weight:
118.94 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

for synthesis

Quality Level

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

bp

333 °C (lit.)

density

4.13 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.[Co++].[O-]C([O-])=O

InChI

1S/CH2O3.Co.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-2

InChI key

JLAIPADPFFTYLP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
Kaluza L, et al.
Applied Catalysis. B, Environmental, 162, 430-436 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service