Skip to Content
Merck
All Photos(2)

Key Documents

537462

Sigma-Aldrich

Isopropyl acetate

≥99.6%

Synonym(s):

IPrOAc, Acetic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740761
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.6%
bp:
85-91 °C (lit.)
vapor pressure:
47 mmHg ( 20 °C)

vapor density

3.5 (vs air)

Quality Level

vapor pressure

47 mmHg ( 20 °C)

Assay

≥99.6%

form

liquid

autoignition temp.

894 °F

expl. lim.

1.8 %, 37 °F
8 %

impurities

≤0.005% Acetic acid
≤0.1% Water
≤0.2% 2-Propanol

refractive index

n20/D 1.377 (lit.)

bp

85-91 °C (lit.)

mp

−73 °C (lit.)

solubility

water: soluble 31.9 g/L at 20 °C

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)=O

InChI

1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3

InChI key

JMMWKPVZQRWMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isopropyl acetate may be employed as a model oxygenate compound to evaluate the catalytic efficiency of La0.8Sr0.2MnO3+x perovskite catalyst for the oxidation of various oxy-derivative compounds. It may be used as an extracting reagent for the N,N-dimethyl-2-[5-(cyanomethyl)-1H-indol-3-yl]ethylamine.

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heats of combustion and formation of ethyl acetate and isopropyl acetate.
Butwill ME and Rockenfeller JD.
Thermochimica Acta, 1(3), 289-295 (1970)
A theoretical study of the pyrolysis of isopropyl acetate.
Silva AM.
Chemical Physics Letters, 439(1), 8-13 (2007)
Vapor-liquid equilibria of carbon dioxide with isopropyl acetate, diethyl carbonate and ethyl butyrate at elevated pressures.
Cheng CH and Chen YP.
Fluid Phase Equilibria, 234(1), 77-83 (2005)
Madan Lal Verma et al.
Enzyme research, 2011, 919386-919386 (2011-05-24)
Selective production of fragrance fatty acid ester from isopropanol and acetic acid has been achieved using silica-immobilized lipase of Bacillus cereus MTCC 8372. A purified thermoalkalophilic extracellular lipase was immobilized by adsorption onto the silica. The effects of various parameters
Improved Fischer indole reaction for the preparation of N,N-dimethyltryptamines: Synthesis of L-695,894, a potent 5-HT1D receptor agonist.
Chen CY, et al.
The Journal of Organic Chemistry, 59(13), 3738-3741 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service