Skip to Content
Merck
All Photos(1)

Documents

13138

Sigma-Aldrich

Magnesium oxide

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

description

filling volume <= 0.15 g/mL

Quality Level

grade

puriss.

Assay

98.0-100.5% (calc. for dried substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E 530

reaction suitability

reagent type: catalyst
core: magnesium

impurities

free alkali, in accordance
residual solvents, complies
soluble salts, in accordance
≤0.002% heavy metals (as Pb)
≤0.1% insoluble in HCl
≤0.1% insoluble in acetic acid
≤1% water-soluble matter

loss

≤5% loss on ignition

mp

2852 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤1%
Cu: ≤10 mg/kg
Fe: ≤200 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

passes test for appearance of solution

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium oxide (MgO) has been reported as an industrially important high-temperature material. Lithium-doped magnesium oxide (Li/MgO) may be employed for the synthesis of ethylene and ethane from methane. It participates as a catalyst for the cycloaddition of carbon dioxide to epoxide. It plays the role of a catalyst during the reaction of carbon dioxide with (R)-styrene to afford (R)-phenyl carbonate.
Magnesium oxide can be used as a wide-band-gap insulator. It can undergo hydration to give magnesium hydroxide nanoparticles.

Application

Magnesium oxide (MgO) may be employed as a phase stabilizer during the fabrication of porous β-tricalcium phosphate (β-TCP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulk synthesis of single-crystalline magnesium oxide nanotubes
Zhan J, et al.
Inorganic Chemistry, 43(8), 2462-2464 (2004)
Nanoparticles of Mg (OH) 2: synthesis and application to paper conservation
Giorgi R, et al.
Langmuir, 21(18), 8495-8501 (2005)
Synthesis of ethylene and ethane by partial oxidation of methane over lithium-doped magnesium oxide.
Ito T and Lunsford JH.
Nature, 721-722 (1985)
Fabrication of β-TCP foam: Effects of magnesium oxide as phase stabilizer on its properties.
Nikaido T, et al.
Ceramics International, 41(10), 14245-14250 (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 495-495 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service