Skip to Content
Merck
All Photos(2)

Key Documents

45629

Supelco

Erucic acid

analytical standard

Synonym(s):

cis-13-Docosenoic acid, Prifac 2990

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)11COOH
CAS Number:
Molecular Weight:
338.57
Beilstein:
1728049
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

analytical standard

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

358 °C/400 mmHg (lit.)

mp

28-32 °C (lit.)

format

neat

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCCC(O)=O

InChI

1S/C22H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H,23,24)/b10-9-

InChI key

DPUOLQHDNGRHBS-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nian Wang et al.
The New phytologist, 180(4), 751-765 (2008-09-25)
Two ethylmethanesulfonate (EMS) mutant populations of the semi-winter rapeseed cv. Ningyou7 were constructed with high mutant load, to provide a TILLING platform for functional genomics in Brassica napus, and for introduction of novel allelic variation in rapeseed breeding. Forward genetic
Samija Amar et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 116(8), 1051-1061 (2008-03-13)
Improving oil and protein quality for food and feed purposes is an important goal in rapeseed (Brassica napus L.) breeding programs. Rapeseed contains phytosterols, used to enrich food products, and sinapate esters, which are limiting the utilization of rapeseed proteins
Nian Wang et al.
BMC plant biology, 10, 137-137 (2010-07-03)
FAE1 (fatty acid elongase1) is the key gene in the control of erucic acid synthesis in seeds of Brassica species. Due to oil with low erucic acid (LEA) content is essential for human health and not enough LEA resource could
Mukhlesur Rahman et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 117(6), 895-904 (2008-07-18)
A single base change in the Bn-FAE1.1 gene in the A genome and a two-base deletion in the Bn-FAE1.2 gene in the C genome produce the nearly zero content of erucic acid observed in canola. A BAC clone anchoring Bn-FAE1.1
Qi Peng et al.
Plant cell reports, 29(4), 317-325 (2010-02-05)
The fatty acid composition in the seed oil was significantly modified following the introduction of transgenes. To further enhance the desirable characteristics of rapeseed oil, it would be beneficial to develop a new approach for the simultaneous silencing of two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service