Skip to Content
Merck
All Photos(1)

Key Documents

11625

Sigma-Aldrich

Sodium tetraborate decahydrate

puriss., meets analytical specification of Ph. Eur., BP, NF, 99.0-103.0%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Agency

Ph. Eur.

grade

puriss.

Assay

99.0-103.0%

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, NF

impurities

carbonate and Bicarbonate, complies
organic volatile impurities, complies
residual solvents, complies
≤0.001% ammonium (NH4)
≤0.002% heavy metals (as Pb)

pH

9.0-9.6 (20 °C, 5%)

density

1.73 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤3 mg/kg
Ca: ≤100 mg/kg
Fe: ≤20 mg/kg
Mg: ≤10 mg/kg

suitability

complies for appearance of solution

storage temp.

room temp

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingzhou Cui et al.
The Analyst, 134(5), 875-880 (2009-04-22)
We developed a photonic crystal sensing method for diol containing species such as carbohydrates based on a poly(vinyl alcohol) (PVA) hydrogel containing an embedded crystalline colloidal array (CCA). The polymerized CCA (PCCA) diffracts visible light. We show that in the
C Cencetti et al.
Carbohydrate polymers, 90(3), 1362-1370 (2012-09-04)
Silver-loaded dressings are designed to provide the same antimicrobial activity of topical silver, with the advantages of a sustained silver release and a reduced number of dressing changes. Moreover, such type of dressing must provide a moist environment, avoiding fiber
Marco Frasconi et al.
Langmuir : the ACS journal of surfaces and colloids, 25(18), 11097-11104 (2009-08-22)
A highly stable biological film was prepared by casting an aqueous dispersion of protein and composite hydrogel obtained from the polysaccharide Scleroglucan (Sclg) and borax as a cross-linking agent. Heme proteins, such as hemoglobin (Hb), myoglobin (Mb), and horseradish peroxidase
Vijay K Bharti et al.
Tropical animal health and production, 40(2), 111-116 (2008-04-22)
An experiment was undertaken to evaluate the protective role of boron on the serum profile of buffalo calves fed a high fluoride ration. Twelve male Murrah buffalo (Bubalus bubalis) calves of 6-8 months age, divided into three groups of four
Hasan Turkez
Journal of applied toxicology : JAT, 28(5), 658-664 (2007-11-21)
Titanium dioxide (TiO(2)) is a potential carcinogenic/mutagenic agent although it is used in many areas including medical industries and cosmetics. Boron (as boric acid and borax) has also well-described biological effects and therapeutic benefits. In a previous study, sister-chromatid exchanges

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service