Merck
  • Ameliorative effects of boron on serum profile in buffalo (Bubalus bubalis) fed high fluoride ration.
Ameliorative effects of boron on serum profile in buffalo (Bubalus bubalis) fed high fluoride ration.
Tropical animal health and production (2008-04-22)
Vijay K Bharti, Meenakshi Gupta, D Lall
ABSTRACT
An experiment was undertaken to evaluate the protective role of boron on the serum profile of buffalo calves fed a high fluoride ration. Twelve male Murrah buffalo (Bubalus bubalis) calves of 6-8 months age, divided into three groups of four calves in each, were fed basal diets and supplemented with sodium fluoride (NaF, 60 ppm) alone or in combination with borax (Na2B4O7.10H2O, 140 ppm) for 90 days. Boron (B) was added in the ration as borax to make @140 ppm boron (elemental B) on DM basis in treatment II. Dietary F caused a significant (p<0.05) depressing effect on serum Ca and Zn on day 90 which was improved with B supplementation. However, serum Fe and Cu did not show any significant change on F or F+B supplementation. The serum ALP and phosphorus level were increased significantly (p<0.05) on F feeding but declined significantly (p<0.05) when B was fed. The findings suggested beneficial effect of boron on serum minerals and ALP in buffalo calves fed high fluoride ration.
Sigma-Aldrich
Sodium tetraborate decahydrate, JIS special grade, 99.0-103.0%
Sigma-Aldrich
Sodium tetraborate decahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.0-103.0%