Skip to Content
Merck
All Photos(1)

Key Documents

Y0001521

Amoxicillin trihydrate for performance verification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Amoxicillin trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45
Beilstein:
7507120
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amoxicillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O5S.3H2O/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;;;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);3*1H2/t9-,10-,11+,14-;;;/m1.../s1

InChI key

MQXQVCLAUDMCEF-CWLIKTDRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amoxicillin trihydrate for performance verification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of RP-HPLC Method for Simultaneous Estimation of Amoxicillin trihydrate and Flucloxacillin sodium in capsule dosage form.
Nikam.S D, et al.
International Journal of PharmTech Research, 1(3), 935-939 (2009)
Ambrose Agweyu et al.
Tropical medicine & international health : TM & IH, 19(11), 1310-1320 (2014-08-19)
To determine the extent and pattern of treatment failure (TF) among children hospitalised with community-acquired pneumonia at a large tertiary hospital in Kenya. We followed up children aged 2-59 months with WHO-defined severe pneumonia (SP) and very severe pneumonia (VSP)
In acute calculous cholecystitis, antibiotics after cholecystectomy did not reduce infection.
Daniel I Steinberg
Annals of internal medicine, 161(12), JC4-JC4 (2014-12-17)
Gilson C N Franco et al.
International journal of clinical pharmacology and therapeutics, 52(5), 425-430 (2014-04-24)
To compare the pharmacokinetic profiles and to evaluate the bioequivalence of two commercial amoxicillin suspension formulations (500 mg/5 mL AMOXIL®, reference formulation and AMOXI-PED®, test formulation) in healthy Brazilian volunteers. Under fasting condition, 25 volunteers (13 males and 12 females)
Clinical trial evidence to advance the science of cholecystectomy.
Joseph S Solomkin
JAMA, 312(2), 135-136 (2014-07-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service