Skip to Content
Merck
All Photos(1)

Key Documents

33454

Supelco

Potassium clavulanate

VETRANAL®, analytical standard

Synonym(s):

Clavulanate potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8NO5K
CAS Number:
Molecular Weight:
237.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[K+].[H][C@@]12CC(=O)N1[C@@H](C([O-])=O)C(\O2)=C\CO

InChI

1S/C8H9NO5.K/c10-2-1-4-7(8(12)13)9-5(11)3-6(9)14-4;/h1,6-7,10H,2-3H2,(H,12,13);/q;+1/p-1/b4-1-;/t6-,7-;/m1./s1

InChI key

ABVRVIZBZKUTMK-JSYANWSFSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium clavulanate is an oxapenam derivative and belongs to the group of β-lactamase inhibitors. It is used in combination with the penicillin group of antibiotics to give them resistance to β-lactamase.

Application

The analytical standard can be used as follows:
  • Simultaneous analysis of amoxicillin and clavulanate potassium in pharmaceutical formulations by capillary electrophoresis (CE) combined with capacitively coupled contactless conductivity detection (C4D)
  • Development of a stability-indicating high-performance liquid chromatographic (HPLC) method combined with mass spectrometry (MS) to measure amoxicillin, clavulanate potassium, and their impurities in their combined dosage forms
  • Residue analysis of potassium clavulanate and amoxicillin trihydrate by a UV spectrophotometric method in their bulk, combined tablet formulations, and human urine samples
  • HPLC method-based determination of meropenem and potassium clavulanate in the presence of their degradation impurities in intravenous solutions in combination with a diode array detector (DAD)
  • Determination of amoxicillin and clavulanic acid by capillary zone electrophoresis (CZE) in their combined commercial formulations

Biochem/physiol Actions

β-lactamase inhibitor, typically added to amoxicillin to increase its effectiveness.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Resp. Sens. 1 - Self-heat. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Baunoch et al.
Infection and drug resistance, 14, 3275-3286 (2021-08-28)
Studies have shown that multiple genes influence antibiotic susceptibility, but the relationship between genotypic and phenotypic antibiotic susceptibility is unclear. We sought to analyze the concordance between the presence of antibiotic resistance (ABR) genes and antibiotic susceptibility results in urine
Yoram Fleissig et al.
Oral surgery, oral medicine, oral pathology and oral radiology, 113(3), e1-e3 (2012-06-09)
A 58-year-old woman presented to the Oral and Maxillofacial Surgery Clinic experiencing severe limited mouth opening and exposed bone in the socket of the right mandibular third molar 8 months following the extraction of the tooth. The patient had been
Oriol Mitjà et al.
The American journal of tropical medicine and hygiene, 86(6), 1043-1045 (2012-06-06)
We report the first documented case of a mycetoma caused by Nocardia yamanashiensis after the initial description of this species. The 16S-rRNA gene sequence analysis was used to identify the novel species, which showed a similarity of 99.9% to the
Ximena Gonzalo et al.
The Journal of antimicrobial chemotherapy, 68(2), 366-369 (2012-10-17)
To: (i) assess if amoxicillin/clavulanate is a useful option for the management of multidrug-resistant/extensively drug-resistant tuberculosis (MDR/XDR-TB); (ii) assess if meropenem/clavulanate is active against Mycobacterium tuberculosis at concentrations achievable in vivo; and (iii) determine whether there was inhibition of meropenem/clavulanate
B R Smith et al.
Drug intelligence & clinical pharmacy, 19(6), 415-420 (1985-06-01)
Potassium clavulanate is a novel beta-lactamase inhibitor, which, in combination, expands the spectrum of amoxicillin to include many amoxicillin-resistant organisms. Potassium clavulanate is excreted 30-50 percent unchanged renally and its plasma time-course parallels that of amoxicillin. Several studies suggest that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service