Skip to Content
Merck
All Photos(1)

Key Documents

Y0001399

Fulvestrant

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, ICI 182,780, ZD 182780, ZD 9238, ZM 182780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

fulvestrant

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O)ccc24

InChI

1S/C32H47F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-29,38-39H,2-10,13-20H2,1H3/t22-,26-,27+,28+,29-,30+,41?/m1/s1

InChI key

VWUXBMIQPBEWFH-WCCTWKNTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fulvestrant EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Fulvestrant (ICI 182,780) is a selective estrogen receptor down-regulator (SERD). Fulvestrant is a high affinity estrogen receptor antagonist. IC50 = 0.29 nM. Fulvestrant is the first "pure" antiestrogen with no agonistic activity both in vitro and in vivo.
Fulvestrant is a selective estrogen receptor down-regulator (SERD); high affinity estrogen receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Donghai Jiang et al.
Cancer letters, 346(2), 292-299 (2014-01-28)
Drug resistance frequently results in poor prognosis and high 5-year recurrence rate in estrogen receptor-negative (ER-) breast cancer patients. Herein, we examined the reversal effects of fulvestrant on multidrug resistance (MDR) in ER- breast cancer cells. Co-administration of fulvestrant significantly
Yuet-Kin Leung et al.
PloS one, 9(5), e98037-e98037 (2014-05-20)
Fulvestrant (ICI-182,780) has recently been shown to effectively suppress prostate cancer cell growth in vitro and in vivo. But it is unclear whether microRNAs play a role in regulating oncogene expression in fulvestrant-treated prostate cancer. Here, this study reports hsa-miR-765
Stephen Chia et al.
Breast (Edinburgh, Scotland), 17 Suppl 3, S16-S21 (2008-03-21)
With the aromatase inhibitors (AIs) replacing tamoxifen as the first-line treatment for postmenopausal women with hormone receptor-positive early and advanced breast cancer, there is a need to evaluate appropriate endocrine treatment options following AI failure. However, until recently, there were
Nadia Okolowsky et al.
Developmental biology, 391(2), 219-229 (2014-04-29)
Mesenchymal dysplasia (mes) mice harbour a truncation in the C-terminal region of the Hh-ligand receptor, Patched-1 (mPtch1). While the mes variant of mPtch1 binds to Hh-ligands with an affinity similar to that of wild type mPtch1 and appears to normally
Fabrice Journé et al.
Expert opinion on drug safety, 7(3), 241-258 (2008-05-09)
Breast cancer is a major health problem in women of developed Western countries. Whereas estrogen receptor (ER) may be involved in many cases in breast carcinogenesis, its expression in breast tumors may predict a favorable response to hormone therapy. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service