Skip to Content
Merck
All Photos(1)

Key Documents

W385101

Sigma-Aldrich

3-Mercaptohexyl acetate

≥98%, FG

Synonym(s):

3-sulfanylhexyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2S
CAS Number:
Molecular Weight:
176.28
FEMA Number:
3851
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.234
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity; alliaceous; tropical

SMILES string

CCCC(S)CCOC(C)=O

InChI

1S/C8H16O2S/c1-3-4-8(11)5-6-10-7(2)9/h8,11H,3-6H2,1-2H3

InChI key

JUCARGIKESIVLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F

Flash Point(C)

73 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alvaro Lezaeta et al.
Food research international (Ottawa, Ont.), 108, 595-603 (2018-05-08)
We developed a sensory-based methodology to aromatically enrich wines using different aromatic fractions recovered during fermentations of Sauvignon Blanc must. By means of threshold determination and generic descriptive analysis using a trained sensory panel, the aromatic fractions were characterized, selected
JianCai Zhu et al.
Food chemistry, 245, 775-785 (2017-12-31)
The volatile compounds of mulberries obtained from three cultivars (M. nigra, Y1, M. Macroura, Y2 and M. Alba, Y3) were investigated by gas chromatography-olfactometry (GC-O), odor activity value (OAV), gas chromatography-mass spectrometry (GC-MS) and flame photometric detection (FPD). The results
Toru Kishimoto et al.
Journal of bioscience and bioengineering, 126(3), 330-338 (2018-04-24)
Gas chromatography-olfactometry (GC-O) analyses of the aroma components extracted from Pilsner-type beer were performed. The concentrations of all 76 components, that were identified after the GC-O analyses, were accurately quantitated. The aroma compounds were then used in the following aroma
Xiaotong Lyu et al.
Food chemistry, 346, 128914-128914 (2021-01-09)
The antioxidants sulfur dioxide (50 ppm) and ascorbic acid (100 ppm) were added to grapes soon after harvest at crushing. The chemical composition and sensory profile of Sauvignon Blanc, Pinot Gris and Chardonnay wines were examined, made from grapes collected at three
Leandro Dias Araujo et al.
Food research international (Ottawa, Ont.), 98, 79-86 (2017-06-15)
Elemental sulfur is a fungicide traditionally used to control Powdery Mildew in the production of grapes. The presence of sulfur residues in grape juice has been associated with increased production of hydrogen sulfide during fermentation, which could take part in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service