Skip to Content
Merck
All Photos(2)

Key Documents

706922

Sigma-Aldrich

8-Mercapto-1-octanol

98%

Synonym(s):

8-Hydroxy-1-octanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2(CH2)6CH2OH
CAS Number:
Molecular Weight:
162.29
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.484

density

0.930 g/mL at 25 °C

storage temp.

−20°C

SMILES string

OCCCCCCCCS

InChI

1S/C8H18OS/c9-7-5-3-1-2-4-6-8-10/h9-10H,1-8H2

InChI key

XJTWZETUWHTBTG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salah Alzghoul et al.
Biosensors & bioelectronics, 77, 491-498 (2015-10-13)
Currently used cancer marker for prostate adenocarcinoma (PC), serum prostate-specific antigen (PSA), greatly overestimates PC population. Patients with high PSA levels have to undergo unnecessary but physically painful and expensive procedure such as prostate biopsies repeatedly. The reliability of PC
Zheng Zeng et al.
Physical chemistry chemical physics : PCCP, 19(30), 20101-20109 (2017-07-21)
Carbon nanodots (CNDs) have attracted great attention due to their superior solubility, biocompatibility, tunable photoluminescence, and opto-electronic properties. This work describes a new fluorescence-based spectroelectrochemistry approach to simultaneously study the photoluminescence and wavelength dependent photocurrent of microwave synthesized CNDs. The
Ee Taek Hwang et al.
ChemElectroChem, 4(8), 1959-1968 (2017-09-19)
Coupling light-harvesting semiconducting nanoparticles (NPs) with redox enzymes has been shown to create artificial photosynthetic systems that hold promise for the synthesis of solar fuels. High quantum yields require efficient electron transfer from the nanoparticle to the redox protein, a
Santiago Oviedo-Rouco et al.
Archives of biochemistry and biophysics, 680, 108243-108243 (2020-01-04)
Cytochrome c is a prototypical multifunctional protein that is implicated in a variety of processes that are essential both for sustaining and for terminating cellular life. Typically, alternative functions other than canonical electron transport in the respiratory chain are associated
Mollye Sanders et al.
Biosensors & bioelectronics, 61, 95-101 (2014-05-27)
A miniaturized, localized surface plasmon resonance (LSPR)-coupled fiber-optic (FO) nanoprobe is reported as a biosensor that is capable of label-free, sensitive detection of a cancer protein biomarker, free prostate specific antigen (f-PSA). The biosensor is based on the LSPR at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service