Skip to Content
Merck
All Photos(1)

Key Documents

W358512

Sigma-Aldrich

L-Phenylalanine

99%, natural, FCC, FG

Synonym(s):

(S)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
FEMA Number:
3585
Beilstein:
1910408
EC Number:
MDL number:
UNSPSC Code:
12352209
Flavis number:
17.018
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
corn
food allergen:
no known allergens

biological source

corn

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.320

Assay

99%

mp

270-275 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

COLNVLDHVKWLRT-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service