Skip to Content
Merck
All Photos(1)

Key Documents

93829

Sigma-Aldrich

L-Tyrosine

≥99.0% (NT), suitable for ligand binding assays, BioUltra

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Tyrosine, BioUltra, ≥99.0% (NT)

product line

BioUltra

Assay

≥99.0% (NT)

form

powder or crystals

optical activity

[α]20/D −11.5±1.0°, c = 4% in 1 M HCl

technique(s)

ligand binding assay: suitable

impurities

insoluble matter, passes filter test
≤0.5% foreign amino acids

ign. residue

≤0.1%

loss

≤0.2% loss on drying, 20 °C (HV)

color

white

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: 25 mg/mL, clear, colorless (hot)

anion traces

chloride (Cl-): ≤400 mg/kg
sulfate (SO42-): ≤300 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 310 nm in 1 M HCl at 0.5 M

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Tyrosine was used in tyrosine hydroxylase (TH) enzyme activity assay. It was also used as a substrate for tyrosinase enzyme in the growth media of Vibrio cholerae strains to stimulate melanin formation. It may be used as a precursor in the synthesis of (-)-MY 336a. MY 336a, a β-adrenergic receptor antagonist, was initially isolated from the culture broth of Streptomyces gabonae KY2234 (ATCC 15282). It may be used in the synthesis of 3-bromo-L-tyrosine and 3,5-dibromo-L-tyrosine.

Other Notes

Amino acid precursor of dopamine and other catecholamines
Growth requirements of various microorganisms

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andisheh Eslamboli et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(4), 769-777 (2005-01-28)
The therapeutic potential of glial cell line-derived neurotrophic factor (GDNF) for Parkinson's disease is likely to depend on sustained delivery of the appropriate amount to the target areas. Recombinant adeno-associated viral vectors (rAAVs) expressing GDNF may be a suitable delivery
V E Coyne et al.
Applied and environmental microbiology, 58(9), 2861-2865 (1992-09-01)
Vibrio cholerae synthesized the pigment melanin in response to specific physiological conditions that were stressful to the bacterium. Pigmentation was induced when V. cholerae was subjected to hyperosmotic stress in conjunction with elevated growth temperatures (above 30 degrees C). The
CRC Handbook of Microbiology, 4, 1-1 (1974)
P M Knappskog et al.
Human molecular genetics, 4(7), 1209-1212 (1995-07-01)
Tyrosine hydroxylase (TH) catalyzes the conversion of L-tyrosine to L-dihydroxyphenylalanine (L-DOPA), the rate-limiting step in the biosynthesis of dopamine. Recently, we described a point mutation in hTH (Q381K) in a family of two siblings suffering from progressive L-DOPA-responsive dystonia (DRD)
Robert S Phillips et al.
Amino acids, 44(2), 529-532 (2012-07-28)
L-Tyrosine is converted to 3-bromo-L-tyrosine in good yield by reaction with 1.2 equiv. of DMSO in HBr/AcOH, while reaction with 2.2 equiv. of DMSO under comparable conditions results in formation of 3,5-dibromo-L-tyrosine in good yield. This is the simplest, safest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service