Skip to Content
Merck
All Photos(1)

Documents

W304799

Sigma-Aldrich

α-Terpinyl acetate

FG

Synonym(s):

(±)-α-Terpinyl acetate, predominantly α-isomer, (±)-2-(4-Methyl-3-cyclohexenyl)isopropyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3047
Beilstein:
3198769
EC Number:
Council of Europe no.:
205
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.015
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

refractive index

n20/D 1.465 (lit.)

bp

220 °C (lit.)

density

0.953 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

herbaceous; floral; woody; citrus; spicy

SMILES string

CC(=O)OC(C)(C)C1CCC(C)=CC1

InChI

1S/C12H20O2/c1-9-5-7-11(8-6-9)12(3,4)14-10(2)13/h5,11H,6-8H2,1-4H3

InChI key

IGODOXYLBBXFDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Α-Terpinyl acetate, a monoterpene ester, is a commercially important fragrance molecule. It can be prepared from α-pinene in the presence of H-beta zeolite catalysts. The essential oils obtained from Stachys setifera ssp. iranica, Chamaecyparis obtuse leaves and Thymus willkomii contain Α-terpinyl acetate as one of the main components.

Application


  • Eucalyptus Oils Phytochemical Composition in Correlation with Their Newly Explored Anti-SARS-CoV-2 Potential: in Vitro and in Silico Approaches.: This research evaluates the antiviral potential of Eucalyptus oils, including α-terpinyl acetate, highlighting its role in combating SARS-CoV-2 through various biochemical mechanisms and suggesting its utility in therapeutic formulations (El-Shiekh RA et al., 2024).

  • Systematic analysis of antimicrobial activity, phytochemistry, and in silico molecular interaction of selected essential oils and their formulations from different Indian spices against foodborne bacteria.: Investigates the antimicrobial properties of essential oils containing α-terpinyl acetate, underscoring its effectiveness against foodborne pathogens and its potential in food safety applications (Gupta N et al., 2023).

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conversion of ?-pinene to terpinyl acetate over H-beta zeolites.
Gainsford GJ, et al.
Applied Catalysis A: General, 209(1), 269-277 (2001)
Composition of the essential oil of Stachys setifera CA Mey ssp. iranica growing in Iran.
Javidnia K, et al.
Flavour and Fragrance Journal, 18(4), 299-300 (2003)
Composition and variability of the essential oil of Thymus willkomii.
Phytochemistry, 30(7), 2289-2293 (1991)
Chemical composition and antimicrobial activity of Chamaecyparis obtusa leaf essential oil.
Yang JK, et al.
Fitoterapia, 78(2), 149-152 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service