Skip to Content
Merck
All Photos(4)

Key Documents

S7670

Sigma-Aldrich

Sodium acetate trihydrate

BioXtra, ≥99.0%

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder

autoignition temp.

1112 °F

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

pH

8.5-10 (25 °C, 408 g/L)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium acetate trihydrate has been used:
  • as a component of citrate-acetate buffer for hydroxyproline assay
  • to prepare buffered salt solutions for ultrafiltration experiments
  • to prepare acetate buffer solution for the electrochemical synthesis of indium tin oxide/gold nanoparticles/nickel oxide/poly(Pyrrole-N-propionic acid) (ITO/GNPs/NiO/poly(PPA)) electrode

Biochem/physiol Actions

Sodium acetate trihydrate (SAT), an inorganic salt hydrate has been the focus of numerous scientific studies on its potential as a heat energy storage phase transition material (PCM).

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cartilage tissue engineering
<BIG>Hwang NS, et al.</BIG>
Stem cell assays, 351-373 (2007)
Thermal conductivity enhancement of sodium acetate trihydrate by adding graphite powder and the effect on stability of supercooling
<BIG>Johansen JB, et al.</BIG>
Energy Procedia, 70, 249-256 (2015)
Fouling behavior of zwitterionic membranes: Impact of electrostatic and hydrophobic interactions
<BIG>Hadidi M and Zydney AL</BIG>
Journal of Membrane Science, 452, 97-103 (2014)
Gold nanoparticle/nickel oxide/poly (pyrrole-N-propionic acid) hybrid multilayer film: Electrochemical study and its application in biosensing
<BIG>Karazehir T, et al.</BIG>
Express Polymer Letters, 11 (2017)
Mohd Baasir Gaisawat et al.
Nutrients, 11(9) (2019-08-29)
Probiotic supplementation plays a key role in maintaining intestinal homeostasis due to its ability to modulate gut microbiota. Although their potential as potent antioxidants have previously been explored, their ability to affect the redox status in the gut lumen of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service