![Sodium Acetate Trihydrate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/299/428/a35e3a90-0537-4619-bb08-bf013fea33e0/640/a35e3a90-0537-4619-bb08-bf013fea33e0.png)
Sodium acetate trihydrate
![Sodium Acetate Trihydrate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/299/428/a35e3a90-0537-4619-bb08-bf013fea33e0/640/a35e3a90-0537-4619-bb08-bf013fea33e0.png)
Synonym(s):
Acetic acid sodium salt trihydrate, Sodium acetate trihydrate
Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-18 of 18
Compare
Product Number
Description
Pricing