Skip to Content
Merck
All Photos(1)

Key Documents

PHR1618

Supelco

Miconazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Miconazole, (±)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O
CAS Number:
Molecular Weight:
416.13
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M1880000
traceable to USP 1443409

API family

miconazole

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)C(OCc3c(cc(cc3)Cl)Cl)C[n]2cncc2

InChI

1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2

InChI key

BYBLEWFAAKGYCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Miconazole is an antimycotic-imidazole derivative compound which is utilized for the management of topical fungal infections. It is typically effective against dermatophytes and Candida albicans, and also many Gram-positive germens.[1][2]

Application

Miconazole may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations, and bulk and cream formulations by chromatography[3][1][2] and spectrophotometry[1][4] techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB5528 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of miconazole and econazole in pharmaceutical formulations by derivative UV spectroscopy and liquid chromatography (HPLC)
Cavrini V, et al.
Journal of Pharmaceutical and Biomedical Analysis, 7(12), 1535-1543 (1989)
Miconazole
USP42-NF37
United States Pharmacopeia/National Formulary, 41(5), 2909-2909 (2013)
Development and validation of a simple stability-indicating high performance liquid chromatographic method for the determination of miconazole nitrate in bulk and cream formulations.
De Zan MM, et al.
Talanta, 79(3), 762-767 (2009)
Determination of miconazole in pharmaceutical creams using internal standard and second derivative spectrophotometry.
Wrobel K, et al.
Journal of Pharmaceutical and Biomedical Analysis, 20(1-2), 99-105 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service