Skip to Content
Merck
All Photos(1)

Key Documents

PHL80391

Crocin

phyproof® Reference Substance

Synonym(s):

Crocetin digentiobiose ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H64O24
CAS Number:
Molecular Weight:
976.96
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

CC(=C\C=C\C=C(C)\C=C\C=C(/C)C(=O)O[C@@H]1O[C@H](CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)\C=C\C=C(\C)C(=C)CO[C@@H]3O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O

InChI

1S/C46H68O23/c1-21(12-8-14-23(3)25(5)18-62-43-40(59)36(55)32(51)28(67-43)19-63-44-38(57)34(53)30(49)26(16-47)65-44)10-6-7-11-22(2)13-9-15-24(4)42(61)69-46-41(60)37(56)33(52)29(68-46)20-64-45-39(58)35(54)31(50)27(17-48)66-45/h6-15,26-41,43-60H,5,16-20H2,1-4H3/b7-6+,12-8+,13-9+,21-10-,22-11+,23-14-,24-15+/t26-,27-,28-,29-,30-,31-,32-,33-,34+,35+,36+,37+,38-,39-,40-,41-,43-,44-,45-,46+/m1/s1

InChI key

LUVDBMJRTUBHNX-RSXXRLSLSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

A highly unsaturared diterpene dicarboxylic acid ester found in saffron.
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • The immunoregulatory property of mesenchymal stem cells in Crocin treatment: This research highlights the modulation of inflammatory genes and microRNAs by crocin, suggesting its potential in immunotherapy and pain management strategies (Rostami et al., 2024).

  • Enhancing the purification of crocin-I from saffron: This study explores advanced green solvent techniques for crocin-I extraction, demonstrating significant implications for biochemical analysis and quality control in pharmaceutical research (Hooshyari Ardakani et al., 2024).

  • NAMPT NAD(+) in the senescence delaying effects of saffron in aging mice: This experimental study assesses crocin′s role in anti-aging processes, underlining its pharmacological relevance and potential in geriatric medicine (Xiao et al., 2024).

Biochem/physiol Actions

Crocin alleviates some ethanol-induced impairments of learning and prevents ethanol-induced inhibition of hippocampal long-term potentiation (LTP), a form of activity-dependent synaptic plasticity that may underly learning and memory. Related CNS effects are specific to the digentiobiose ester; crocetin glucose gentiobiose ester is half as potent, and the diglucose ester has no effect at all.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jian Zhang et al.
Annals of translational medicine, 8(18), 1180-1180 (2020-11-27)
Cervical cancer is a prevalent tumor mainly induced by Human Papilloma Virus (HPV). Autophagy was inactivated with HPV to promote cancer progression. Here we explored the effects of crocin on cervical cancer cells, mainly on autophagy and apoptosis. SiHa cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service