Skip to Content
Merck
All Photos(1)

Key Documents

P1061

Supelco

Proadifen hydrochloride

analytical standard, ≥95%

Synonym(s):

α-Phenyl-α-propylbenzeneacetic acid 2-(diethylamino)ethyl ester, N,N-Diethylaminoethyl 2,2-diphenylvalerate, SKF-525A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31NO2 · HCl
CAS Number:
Molecular Weight:
389.96
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

122-123 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl.CCCC(C(=O)OCCN(CC)CC)(c1ccccc1)c2ccccc2

InChI

1S/C23H31NO2.ClH/c1-4-17-23(20-13-9-7-10-14-20,21-15-11-8-12-16-21)22(25)26-19-18-24(5-2)6-3;/h7-16H,4-6,17-19H2,1-3H3;1H

InChI key

FHIKZROVIDCMJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Proadifen hydrochloride may be used as an internal standard in the determination of analgesic agents like O-desmethyltramadol, tramadol and N-desmethyltramadol in human urine samples using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Blocks glibenclamide-sensitive K+ channels. Inhibits neuronal nitric oxide synthase and cytochrome P450

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Fuchs et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158611-158611 (2020-01-10)
Trihydroxyoctadecenoic acids (TriHOMEs) are linoleic acid-derived lipid mediators reported to be dysregulated in obstructive lung disease. In contrast to many other oxylipins, TriHOME biosynthesis in humans is still poorly understood. The association of TriHOMEs with inflammation prompted the current investigation
Simultaneous determination of tramadol, O-desmethyltramadol and N-desmethyltramadol in human urine by gas chromatography?mass spectrometry
El-Sayed Y A-A, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 926, 9-15 (2013)
Amber Woodard et al.
AIMS neuroscience, 6(1), 25-42 (2020-04-29)
Neuronal nitric oxide (nNO) has been shown to affect motor function in the brain. Specifically, nNO acts in part through regulation of dopamine (DA) release, transporter function, and the elicitation of neuroprotection/neurodegeneration of neurons in conditions such as Parkinson's disease
Joseph A Crifasi et al.
Journal of analytical toxicology, 30(8), 581-592 (2006-11-30)
Stir bar sorptive extraction is an innovative sample extraction technique that can be used to process blood, urine, and tissue samples for routine drug screening in the forensic toxicology laboratory. The Gerstel Twister desorption unit (TDU) system is a multifunctional
Daniele Giovanni Ghiglioni et al.
Pharmaceutics, 12(4) (2020-04-25)
In this study, a sensitive quantitative method based on high performance liquid chromatography combined with high-resolution mass spectrometry, Q ExactiveTM-Orbitrap® was set up and applied for the determination of the immunosuppressor agents cyclosporine A and tacrolimus in novel ethanol-free ophthalmic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service