73144
Resorufin
95%
Synonym(s):
7-Hydroxy-3H-phenoxazin-3-one, NSC 12097
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H7NO3
CAS Number:
Molecular Weight:
213.19
Beilstein:
174850
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Quality Level
Assay
≥95.0% (HPLC)
95%
form
solid
mp
>300 °C (lit.)
storage temp.
−20°C
SMILES string
O=C(C=C1)C=C2C1=NC3=CC=C(O)C=C3O2
InChI
1S/C12H7NO3/c14-7-1-3-9-11(5-7)16-12-6-8(15)2-4-10(12)13-9/h1-6,14H
InChI key
HSSLDCABUXLXKM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Red fluorophore standard. Widely used in enzyme assay systems. Absorption and fluorescence of resorufin are pH dependent. Below the pKa (~6.0), Abs. shifts to ~ 480 nm and both E and fluorescence quantum yield are markedly lower. Resorufin is unstable in the presence of thiols such as dithiothreitol (DTT) and 2-mercaptoethanol.
Resorufin has high extinction coefficient and good chemical and photostability. These properties of resorufin make it perfect for its usage in inverse fluorescence assay to detect H2O2 by measuring the disappearance of its fluorescence upon horseradish peroxidase-mediated oxidation to a nonfluorescent derivative.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A stable nonfluorescent derivative of resorufin for the fluorometric determination of trace hydrogen peroxide: applications in detecting the activity of phagocyte NADPH oxidase and other oxidases.
Zhou M, et al.
Analytical Biochemistry, 253(2), 162-168 (1997)
Matteo Miceli et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-19)
Monoacylglycerol lipase (MAGL) is a serine hydrolase that has a key regulatory role in controlling the levels of 2-arachidonoylglycerol (2-AG), the main signaling molecule in the endocannabinoid system. Identification of selective modulators of MAGL enables both to provide new tools
Ahmed A El-Sherbeni et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1498-1507 (2014-06-28)
Cytochrome P450 (P450) enzymes mediate arachidonic acid (AA) oxidation to several biologically active metabolites. Our aims in this study were to characterize AA metabolism by different recombinant rat P450 enzymes and to identify new targets for modulating P450-AA metabolism in
David Hess et al.
Analytical chemistry, 87(9), 4965-4972 (2015-04-08)
Droplet-based microfluidic systems offer a range of advantageous features for the investigation of enzyme kinetics, including high time resolution and the ability to probe extremely large numbers of discrete reactions while consuming low sample volumes. Kinetic measurements within droplet-based microfluidic
Julian G Jude et al.
Oncogene, 34(10), 1253-1262 (2014-04-01)
Given the importance of deregulated phosphoinositide (PI) signaling in leukemic hematopoiesis, genes coding for proteins that regulate PI metabolism may have significant and as yet unappreciated roles in leukemia. We performed a targeted knockdown (KD) screen of PI modulator genes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service