Skip to Content
Merck
All Photos(2)

Key Documents

N8280

Sigma-Aldrich

Naproxen

meets USP testing specifications

Synonym(s):

(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Beilstein:
3591068
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

powder

mp

152-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Haupt et al.
Journal of biochemical and biophysical methods, 25(4), 273-284 (1992-12-01)
A column-switching system for fast determination of (R)- and (S)-naproxen in liver microsomes has been developed. The centrifuged sample was injected directly onto a pre-column with octadecylcoated silica. The retained analytes were then directed to an alpha 1-AGP column using
Jiayang Li et al.
Journal of the American Chemical Society, 135(2), 542-545 (2012-11-10)
As systemically used therapeutics for treating acute or chronic pains or inflammations, nonsteroidal anti-inflammatory drugs (NSAIDs) also associate with the adverse gastrointestinal and renal effects and cardiovascular risks. Thus, it is beneficial to develop topical gels that selectively inhibit cyclooxygenase-2
J Rossat et al.
Clinical pharmacology and therapeutics, 66(1), 76-84 (1999-08-03)
To compare the renal hemodynamic and tubular effects of celecoxib, a selective inhibitor of cyclooxygenase-2 (COX-2) to those of naproxen, a nonselective inhibitor of cyclooxygenases in salt-depleted subjects. Forty subjects were randomized into four parallel groups to receive 200 mg
Michela Campolo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(11), 4489-4499 (2013-08-01)
Spinal cord trauma causes loss of motor function that is in part due to the ensuing inflammatory response. Hydrogen sulfide (H2S) is a potent, endogenous anti-inflammatory and neuroprotective substance that has been explored for use in the design of novel
Dong Qing Zhang et al.
Chemosphere, 91(1), 14-21 (2012-12-27)
Scirpus validus was grown hydroponically and exposed to the pharmaceuticals, carbamazepine and naproxen at concentrations of 0.5-2.0 mg L(-1) for an exposure duration of up to 21 d. By the end of experiment, carbamazepine elimination from the nutrient solution reached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service