Skip to Content
Merck
All Photos(3)

Documents

N5160

Sigma-Aldrich

Naproxen sodium

meets USP testing specifications

Synonym(s):

(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NaO3
CAS Number:
Molecular Weight:
252.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

powder

application(s)

pharmaceutical (small molecule)

SMILES string

[Na+].COc1ccc2cc(ccc2c1)[C@H](C)C([O-])=O

InChI

1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1

InChI key

CDBRNDSHEYLDJV-FVGYRXGTSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cyclooxygenase (Prostaglandin H synthase 1 and 2) inhibitor.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arın Gül Dal et al.
Journal of analytical methods in chemistry, 2014, 352698-352698 (2014-10-09)
Simple and rapid capillary zone electrophoretic method was developed and validated in this study for the determination of piroxicam in tablets. The separation of piroxicam was conducted in a fused-silica capillary by using 10 mM borate buffer (pH 9.0) containing 10%
Rory W Blackler et al.
American journal of physiology. Gastrointestinal and liver physiology, 308(12), G994-1003 (2015-04-18)
The small intestine is a significant site of ulceration and bleeding induced by nonsteroidal anti-inflammatory drugs (NSAIDs). The pathogenesis is poorly understood. The present study explored the roles of bile, bacteria, and enterohepatic circulation to NSAID enteropathy, using both a
Jason A Miranda et al.
PloS one, 9(8), e106108-e106108 (2014-08-27)
Sensory processing in the spinal cord during disease states can reveal mechanisms for novel treatments, yet very little is known about pain processing at this level in the most commonly used animal models of articular pain. Here we report a
Matteo Morotti et al.
European journal of obstetrics, gynecology, and reproductive biology, 179, 63-68 (2014-06-27)
Evaluate patient satisfaction at 6-month treatment in women with symptomatic rectovaginal endometriosis and migraine without aura with (progestogen-only contraceptive pill, POP versus sequential combined oral contraceptives, COC) STUDY DESIGN: A patient preference trial including 144 women (82 in the group
Tarjinder Sahota et al.
Toxicology and applied pharmacology, 278(3), 209-219 (2014-03-29)
The assessment of safety in traditional toxicology protocols relies on evidence arising from observed adverse events (AEs) in animals and on establishing their correlation with different measures of drug exposure (e.g., Cmax and AUC). Such correlations, however, ignore the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service