Skip to Content
Merck
All Photos(2)

Key Documents

N5410

Sigma-Aldrich

Nicotinic acid

meets USP testing specifications

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

99.0-101.0% dry basis

form

powder

color

white

mp

236-239 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)
rat ... Chrnb2(54239)

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid, also referred to as niacin or as vitamin B3, is a water-soluble vitamin. Nicotinic acid is synthesized from the amino acid tryptophan and is present majorly in peanuts, fish and lean meat.

Application

Nicotinic acid has been used as an antioxidant to evaluate its protective functionality in colon LoVo cell lines against doxorubicin induced cytotoxicity.

Biochem/physiol Actions

Nicotinic modulates lipid metabolism and may be useful in treating dyslipidemia. It is an antioxidant and acts as a coenzyme in the form of nicotinamide adenine nucleotides. Deficiency of niacin leads to enhanced lipid peroxidation and is also implicated in Crohn′s disease Deficiency also impacts DNA repair and also leads to skin and gastrointestinal disorder, pellagra. Nicotinic acid reduces the low-density lipoprotein (LDL) synthesis and improves high-density lipoprotein (HDL) levels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nicotinic acid: a lipid-lowering agent with unrealized potential
Aboulsoud SH
The Egyptian Journal of Internal Medicine, 26(1), 1-1 (2014)
Pellagra: a non-eradicated old disease
Savvidou S
Clinics and practice, 4(1) (2014)
Niacin deficiency delays DNA excision repair and increases spontaneous and nitrosourea-induced chromosomal instability in rat bone marrow
Kostecki LM
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 625(1-2), 50-61 (2007)
Nicotinic acid: an old drug with a promising future
Bodor ET and Offermanns S
British Journal of Pharmacology, 153(S1), S68-S75 (2008)
Effect of pO2 on antitumor drug cytotoxicity on MDR and non-MDR variants selected from the LoVo metastatic colon carcinoma cell line
Lelong-Rebel I, et al.
Anticancer Research, 28(1A), 55-68 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service