Skip to Content
Merck
All Photos(1)

Documents

M5260

Supelco

(+)-Methamphetamine hydrochloride solution

analytical standard, for drug analysis, 1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N · HCl
CAS Number:
Molecular Weight:
185.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CN[C@@H](C)Cc1ccccc1

InChI

1S/C10H15N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h3-7,9,11H,8H2,1-2H3;1H/t9-;/m0./s1

InChI key

TWXDDNPPQUTEOV-FVGYRXGTSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer E Naylor et al.
Drug and alcohol dependence, 149, 280-284 (2015-02-25)
Synthetic cathinones are beta-ketophenethylamine analogs manufactured to avoid legal restrictions placed on illicit stimulants like methamphetamine. Regulating these "emerging" designer drugs require scientific evidence of abuse potential. The present study evaluated the discriminative-stimulus effects of three synthetic cathinones, recently identified
Shaolin Yang et al.
Addiction biology, 20(1), 69-79 (2013-08-06)
(1) H magnetic resonance spectroscopy has demonstrated alterations in several neurometabolites in methamphetamine (METH)-dependent individuals in brain regions implicated in addiction. Yet, it is unclear whether these neurochemicals return to homeostatic levels after an individual abstains from drug use, a
Emily C Eastwood et al.
Behavioural brain research, 272, 111-120 (2014-07-01)
Genetic factors likely influence individual sensitivity to positive and negative effects of methamphetamine (MA) and risk for MA dependence. Genetic influence on MA consumption has been confirmed by selectively breeding mouse lines to consume high (MAHDR) or low (MALDR) amounts
Betina González et al.
Neuropharmacology, 87, 188-197 (2014-02-18)
Chronic use of methamphetamine (METH) leads to long-lasting cognitive dysfunction in humans and in animal models. Modafinil is a wake-promoting compound approved for the treatment of sleeping disorders. It is also prescribed off label to treat METH dependence. In the
Joseph A Pipkin et al.
Drug and alcohol dependence, 142, 341-344 (2014-07-22)
Nicotine is commonly abused in adolescence and is believed to be a "gateway" to other drugs of abuse [e.g., methamphetamine (METH)]. The relationship between early nicotine exposure and later METH use is complicated because the majority of juvenile smokers continue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service