M2178
Mycoplasma agar base
suitable for microbiology, NutriSelect® Plus
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.8±0.2 (25 °C)
application(s)
clinical testing
food and beverages
life science and biopharma
veterinary
microbiology
suitability
mycoplasma
Application
Used for isolation and cultivation of mycoplasma.
Components
Ingredients (g/L)
Beef heart, infusion, 250.00
Peptic digest of animal tissue, 10.00
Sodium chloride, 5.00
Agar, 15.00
Beef heart, infusion, 250.00
Peptic digest of animal tissue, 10.00
Sodium chloride, 5.00
Agar, 15.00
Preparation Note
Suspend 36 g of Mycoplasma Agar Base in 700 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 45°C and aseptically add a 300 ml 1% solution of Horse Serum to the agar base or add 10 vials of Mycoplasma Enrichment Supplement (Cat. No. M6930) to the Mycoplasma Agar Base. Mix well before dispensing. Dispense into petri plates or tubes.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service