Skip to Content
Merck
All Photos(1)

Documents

H1270

Sigma-Aldrich

Horse Serum

Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

HS, equine sera, equine serum, horse sera, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

horse serum

sterility

sterile-filtered

form

liquid

composition

Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

≤10 EU/mL Endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Horse serum produced from adult horses is homogenous and has low levels of polyamine oxidase. It is metabolically stable and promotes cell proliferation. Horse sera is an alternate to bovine serum and is a source of growth factors and hormones.

Application

Horse Serum has been used as a supplement in culture medium in pheochromocytoma cell culture, cortical slice cultures, retinal cells and myotubes.

Other Notes

Donor Horse Serum is collected from controlled donor herds located in the USA. Horse serum contains high protein and low trace metals compared to Fetal Bovine Serum. Donor Horse Serum is suitable for use in diagnostic assays, as a supplement in mycoplasma growth and assay media and for culturing hematopoietic stem cells and neuronal cells.

Preparation Note

Collected from a controlled herd.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Z Kubicek-Sutherland et al.
Toxins, 11(4) (2019-04-17)
Mycolactone, the amphiphilic macrolide toxin secreted by Mycobacterium ulcerans, plays a significant role in the pathology and manifestations of Buruli ulcer (BU). Consequently, it follows that the toxin is a suitable target for the development of diagnostics and therapeutics for
Myostatin induces atrophy of trout myotubes through inhibiting the TORC1 signaling and promoting Ubiquitin-Proteasome and Autophagy-Lysosome degradative pathways
Seiliez I, et al.
General and Comparative Endocrinology, 186, 9-15 (2013)
Anna T Bajur et al.
Journal of cell science, 132(15) (2019-07-14)
During morphogenesis, epithelia undergo dynamic rearrangements, which requires continuous remodelling of junctions and cell shape, but at the same time mechanisms preserving cell polarity and tissue integrity. Apico-basal polarity is key for the localisation of the machinery that enables cell
Patterning cultured cells by visible light illumination with photosensitizers
Waki R, et al.
Journal of Bioscience and Bioengineering, 100(3), 331-334 (2005)
Mechanical spectroscopy of retina explants at the protein level employing nanostructured scaffolds
Rahman SM, et al.
Soft Matter, 12(14), 3431-3441 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service