Skip to Content
Merck
All Photos(1)

Key Documents

M0350000

Menthol

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

menthol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC1C(CCC(C1)C)C(C)C

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Menthol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodney B Croteau et al.
Die Naturwissenschaften, 92(12), 562-577 (2005-11-18)
(-)-Menthol is the most familiar of the monoterpenes as both a pure natural product and as the principal and characteristic constituent of the essential oil of peppermint (Mentha x piperita). In this paper, we review the biosynthesis and molecular genetics
Joseph A Farco et al.
Mini reviews in medicinal chemistry, 13(1), 124-131 (2012-10-16)
Menthol, a natural product of the peppermint plant Mentha x piperita (Lamiaceae), is a monoterpene which is widely used as a natural product in cosmetics, a flavoring agent, and as an intermediate in the production of other compounds. Various extracts
Jennifer M Kreslake et al.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco, 10(4), 705-715 (2008-04-18)
The use of menthol in cigarettes is actively promoted by the tobacco industry for its perceived sensory benefits, and smokers of menthol cigarettes commonly differ from nonmenthol smokers in markers of smoking behavior and addiction. In this study, we analyzed
Stacey J Anderson
Tobacco control, 20 Suppl 2, ii20-ii28 (2011-04-29)
To examine tobacco industry marketing of menthol cigarettes and to determine what the tobacco industry knew about consumer perceptions of menthol. A snowball sampling design was used to systematically search the Legacy Tobacco Documents Library (LTDL) (http://legacy.library.ucsf.edu) between 28 February
Jonathan Foulds et al.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco, 12 Suppl 2, S102-S109 (2011-01-05)
Menthol cigarette smokers may find it harder to quit smoking than smokers of nonmenthol cigarettes. We conducted a systematic review of published studies examining the association between menthol cigarette smoking and cessation. Electronic databases and reference lists were searched to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service