Skip to Content
Merck
All Photos(1)

Documents

PHR1214

Supelco

Methyl salicylate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1537450

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

API family

methyl salicylate

CofA

current certificate can be downloaded

autoignition temp.

847 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Methyl salicylate is one of the common ingredients present in topical analgesics and rubifacients. It is prescribed for the management of minor muscle aches and pains.

Application

Methyl salicylate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and voltammetry techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3556 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fourier transform mid-infrared (MIR) and near-infrared (NIR) spectroscopy for rapid quality assessment of Chinese medicine preparation Honghua Oil
Wu YW, et al.
Journal of Pharmaceutical and Biomedical Analysis, 46(3), 498-504 (2008)
Determination of salicylic acid in pharmaceutical formulations and foods by differential-pulse voltammetry using a glassy carbon electrode
Fung YS and Luk SF
Analyst, 114(8), 943-945 (1989)
Optimization and validation of liquid chromatography and headspace-gas chromatography based methods for the quantitative determination of capsaicinoids, salicylic acid, glycol monosalicylate, methyl salicylate, ethyl salicylate, camphor and l-menthol in a topical formulation
Pauwels J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 60(3), 51-58 (2012)
Agata Bartyzel et al.
Analytica chimica acta, 707(1-2), 204-209 (2011-10-27)
N,N'-bis-(α-methylsalicylidene)-2,2-dimethyl-1,3-propanediimine (SBTD) modified silica gel was prepared and used as sorbent for solid phase extraction of Cr(III) ions from aqueous solution. This sorbent showed a high sorption affinity for Cr(III) while recovery of Cr(VI) was very low. The analyte ion
Hsu Shao Feng et al.
Journal of clinical periodontology, 38(7), 637-643 (2011-04-01)
To evaluate the clinical efficacy of subgingival ultrasonic instrumentation irrigated with essential oils (EOs) of residual periodontal pockets. Sixty-four individuals with chronic periodontitis were invited to participate in this randomized, double-blind, parallel, and placebo-controlled clinical trial. All subjects received non-surgical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service