All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H22N2O2
Molecular Weight:
346.42
UNSPSC Code:
12352101
NACRES:
NA.22
Recommended Products
Assay
≥98% (HPLC)
form
powder
storage temp.
2-8°C
Application
Fluorescence microscopy, fluorescence spectroscopy, conjugation.
Features and Benefits
LightOx72 is a fluorescent molecule that exhibits solvatochromatic fluorescence emission when excited with UV-A and violet (405 nm) light sources. In the free, unconjugated form LightOx72 can be used as a cellular membrane stain. The free secondary amine moiety is a reactive group for conjugation to other (bio)molecules by N-acylation, N-sulphonylation, N-alkylation or aza-Michael addition. Stock solutions are typically prepared at 10 mM in DMSO or DMF.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Other Notes
Chemical Equivalency 1ea=1mg
Legal Information
LightOx is a trademark of LightOx Ltd.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service