Skip to Content
Merck
All Photos(1)

Key Documents

LIGHTOX21

Sigma-Aldrich

LightOx21

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO2
CAS Number:
Molecular Weight:
319.40
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

N1(CCC(c2c1ccc(c2)C#Cc3ccc(cc3)C(=O)O)(C)C)C

InChI key

WQRGZQYIDRZUOE-UHFFFAOYSA-N

Application

Fluorescence microscopy, fluorescent retinoid binding assay, fluorescence spectroscopy.

Biochem/physiol Actions

Strong ligand for retinoic acid receptor proteins

Features and Benefits

LightOx21 is a synthetic analogue of all-trans-retinoic acid (ATRA), that exhibits solvatochromatic fluorescence emission when excited with UV or 405 nm light. The compound is a strong ligand for the retinoic acid receptor proteins (RARs) and related cellular retinoic acid binding proteins (CRABPI/II) and can, therefore, be used as a replacement for ATRA in cellular assays involving retinoid signaling processes. LightOx21 can be utilised to study retinoid behaviour using fluorescence microscopy, employed in a competitive fluorometric ligand-binding assay for direct determination of the strength of binding of non-fluorescent retinoids to a protein target, and as a fluorophore in fluorescence spectroscopy.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Other Notes

Chemical Equivalency 1ea=1mg

Legal Information

LightOx is a trademark of LightOx Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service