Skip to Content
Merck
All Photos(1)

Documents

C4206

Supelco

Carbamazepine 10,11-epoxide

analytical standard

Synonym(s):

1a,10b-Dihydro-6H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

NC(=O)N1c2ccccc2C3OC3c4ccccc14

InChI

1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)

InChI key

ZRWWEEVEIOGMMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbamazepine 10,11-epoxide is a metabolite of the drug carbamazepine.

Application

Carbamazepine 10,11-epoxide may be used as a test material in the quantification of histone deacetylases(HDAC) inhibition using fluorescent HDAC activity assay.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

First metabolite of carbamazepine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciana Vera-Candioti et al.
Electrophoresis, 29(22), 4527-4537 (2008-11-28)
Drug monitoring in serum samples was performed using second-order data generated by CE-DAD, processed with a suitable chemometric strategy. Carbamazepine could be accurately quantitated in the presence of its main metabolite (carbamazepine epoxide), other therapeutic drugs (lamotrigine, phenobarbital, phenytoin, phenylephrine
Ying-Chang Chi et al.
Toxicology and applied pharmacology, 263(3), 315-322 (2012-07-21)
Carbamazepine (CBZ), an antiepileptic with narrow therapeutic window, is a substrate of CYP 3A which metabolizes CBZ to carbamazepine-10,11-epoxide (CBZE), an active metabolite. This study investigated the acute and chronic effects of Polygonum cuspidatum (PC), a resveratrol-rich nutraceutical, on the
L Bertilsson et al.
Clinical pharmacokinetics, 11(3), 177-198 (1986-05-01)
Carbamazepine is a first-line drug in the treatment of most forms of epilepsy and also the drug of first choice in trigeminal neuralgia. Furthermore, it is now frequently used in bipolar depression. Most oral formulations of carbamazepine are well absorbed
Ping Ji et al.
Journal of clinical pharmacology, 48(8), 948-956 (2008-06-04)
The effect of efavirenz on the pharmacokinetics of carbamazepine and vice versa was investigated in adult healthy subjects in a randomized, open-label, 2-period crossover, multiple-dose study. Subjects were randomized to receive either efavirenz 600 mg qd for 14 days or
G Bellucci et al.
Journal of medicinal chemistry, 30(5), 768-773 (1987-05-01)
Carbamazepine 10,11-oxide (1a,10b-dihydro-6H-dibenzo[b,f]oxireno[d]azepine-6-carboxamide), a key intermediate in carbamazepine metabolism, was found to be unusually resistant to enzymatic hydrolysis when incubated with microsomal and cytosolic fractions from rabbit, rat, and guinea pig livers. However, its hydrolysis product, trans-10,11-dihydro-10,11-dihydroxy-5H-dibenzo[b,f]azepine-5-carboxamide , was excreted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service