Skip to Content
Merck
All Photos(3)

Documents

T9250

Sigma-Aldrich

Thymidine

≥99%

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Beilstein:
89285
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99%

form

powder

mp

186-188 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1

InChI key

IQFYYKKMVGJFEH-XLPZGREQSA-N

Gene Information

human ... DTYMK(1841)

Looking for similar products? Visit Product Comparison Guide

General description

Thymidine is also referred to as pyrimidine deoxynucleoside. Deoxythymidine is a nucleoside present in DNA. In a DNA double stranded structure, thymidine pairs with deoxyadeninosine. It is useful in cell synchronization during S-phase. In the salvage pathway of pyrimidines, pyrimidine phosphorylase reversibly converts thymine to thymidine.

Application

Thymidine has been used:
  • as a supplement in HL5-modified medium for cell culture
  • to incubate cells in cell-cycle analysis
  • in cell synchronization

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molecular Biology, Principles of Genome Function: Biology, Molecular biology (2016)
Kif20a inhibition reduces migration and invasion of pancreatic cancer cells
Stangel D, et al.
The Journal of Surgical Research, 197(1), 91-100 (2015)
Phosphorylation of Ku dictates DNA double-strand break (DSB) repair pathway choice in S phase
Lee KJ, et al.
Nucleic Acids Research, 44(4), 1732-1745 (2015)
Dominique Desbouis et al.
Journal of medicinal chemistry, 51(21), 6689-6698 (2008-10-08)
Human cytosolic thymidine kinase (hTK1) has proven to be a suitable target for noninvasive imaging of cancer cell proliferation using radiolabeled substrates such as [ (18)F]fluorothymidine ([ (18)F]FLT). However, a thymidine tracer useful for single photon emission tomography (SPECT) based
Human Biochemistry and Disease, 571-571 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service