Skip to Content
Merck
All Photos(1)

Key Documents

BCR050

Benzo[e]pyrene

BCR®, certified reference material

Synonym(s):

1,2-Benzpyrene, 4,5-Benzpyrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1911334
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

177-180 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)c3cccc4ccc5cccc2c5c34

InChI

1S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12H

InChI key

TXVHTIQJNYSSKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR050

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W H Houser et al.
Molecular carcinogenesis, 5(3), 232-237 (1992-01-01)
In the rat, expression of the CYP1A1 gene is closely associated with arylhydrocarbon hydroxylase (AHH) enzyme activity. AHH is an inducile enzyme activity known to play an important role in the bioactivation of polycyclic aromatic hydrocarbons (PAHs) to mutagenic and
Saffar Mansoor et al.
Investigative ophthalmology & visual science, 51(5), 2601-2607 (2009-12-05)
To study the inhibitory effects of some agents or drugs (inhibitors) on benzo(e)pyrene (B(e)P)-induced cell death and apoptosis on human retinal pigment epithelial (ARPE-19) cells in vitro. ARPE-19 cells were pretreated with varying concentrations of different classes of inhibitors (calpain
Silvina Fioressi et al.
Environmental science & technology, 39(10), 3646-3655 (2005-06-16)
The photodegradation of benzo[e]pyrene (BeP), a ubiquitous polycyclic aromatic hydrocarbon (PAH) contaminant, was investigated in solution and adsorbed on surfaces modeling the atmospheric particulate matter to provide fundamental information that could help to clarify its fate in the atmosphere. Diones
Joris J H Haftka et al.
Environmental toxicology and chemistry, 27(7), 1526-1532 (2008-02-12)
The uptake kinetics of fluorene, phenanthrene, fluoranthene, pyrene, and benzo[e]pyrene by solid-phase microextraction fibers was studied in the presence of dissolved organic matter (DOM) obtained from sediment pore water and resulted in increased fiber absorption and desorption rate coefficients. Compared
D W Rosenberg et al.
Archives of biochemistry and biophysics, 300(1), 186-192 (1993-01-01)
The consequences of altered cytochrome P450-dependent monooxygenase activities in colonic tissue are unknown. As an initial step toward elucidating underlying mechanisms that regulate cytochrome P450 levels in colonic epithelium, we have characterized CYP1A1(3) induction in cultured human colonic cells (Caco-2).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service