B3883
Bile bovine
dried, unfractionated
Synonym(s):
Ox gall powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
dried, unfractionated
color
yellow-brown
solubility
water: soluble 50 mg/mL
application(s)
microbiology
General description
Bile bovine is extracted under controlled conditions from purified fresh bile, to be used in bacteriological culture media as a selective inhibitory agent for isolation and identification of pathogens. Bile is a yellow-green aqueous solution consisting of 70% bile salts, 22% phospholipids, 4% cholesterol, 3% proteins and 0.3% bilirubin. It is produced from cholesterol in liver hepatocytes and stored in the gallbladder. Bile acids are steroid acids found mainly in the bile of mammals and other vertebrates. They can be used as a selective agent in media for the detection of intestinal bacteria.
Application
Bile bovine has been used:
- in De Man, Rogosa and Sharpe agar (MRS) broth for the growth test under high bile-salt condition
- as a component in MRS to grow Lactobacillus delbrueckii subsp. lactis 200+,to study the effect of bile salts on cell morphology
- as a component in brain heart infusion (BHI) broth for broth assays to study its effects on Enterococcus faecalis V583
Biochem/physiol Actions
Bile acids plays a physiological role in the removal of cholesterol from the body. It also participates in the intestinal solubilization and absorption of lipids.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Screening lactic acid bacteria from swine origins for multistrain probiotics based on in vitro functional properties
Anaerobe, 16(4), 321- 326 (2010)
Bile acid patterns in commercially available oxgall powders used for the evaluation of the bile tolerance ability of potential probiotics
PLoS ONE, 13(3), e0192964-e0192964 (2018)
Transcriptional responses of Enterococcus faecalis V583 to bovine bile and sodium dodecyl sulfate
Applied and Environmental Microbiology, 73(18), 5767- 5774 (2007)
Food research international (Ottawa, Ont.), 94, 45-53 (2017-03-16)
Malabsorption of dietary sugars is a common cause of gastrointestinal discomfort, affecting up to one in three people with debilitating symptoms, such as abdominal pain, osmotic diarrhoea, bloating and flatulence. Besides dietary interventions, it has been suggested that ingestion of
Impact of bile salt adaptation of Lactobacillus delbrueckii subsp. lactis 200 on its interaction capacity with the gut
Research in Microbiology, 162(8), 782- 790 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service