Skip to Content
Merck
All Photos(2)

Documents

48305

Millipore

Bile salts

suitable for microbiology

Synonym(s):

Bile acids sodium salt, cholic acid-deoxycholic acid sodium salt mixture, Bile acids sodium salt, Cholic acid-Deoxycholic acid sodium salt mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

description

anionic

Quality Level

sterility

non-sterile

Assay

40-60% (Cholic Acid ; approximately 50% sodium cholate and approximately 50% sodium deoxycholate, HPLC)

form

powder

composition

cholic acid sodium salt, ~50%
deoxycholic acid sodium salt, ~50%

impurities

≤5% water

loss

≤5% loss on drying (water)

solubility

H2O: 0.1 g/mL, clear, faintly yellow

application(s)

microbiology

storage temp.

10-30°C

General description

Bile salt is an organic sodium salt with a conjugate of any of the bile acids along with either glycine or taurine. Biles salts are composed of four different kinds of bile acids - cholic, deoxycholic, chenodeoxycholic and lithocholic acids.
Bile salts are extracted under controlled conditions from purified fresh bile, to be used in bacteriological culture media, where it behaves as a selective inhibitory agent that enables isolation and identification of pathogens. For example, in MacConkey Agar/Broth.

Application

Bile salts may be used as one of the selective agents to prevent the growth of injured Escherichia coli cells, in an experiment done in order to identify the specific agent in the selective media which would inhibit the growth of the injured Escherichia coli ML30 cells. It may also be used for identification of isolates of Vibrio (Beneckea) vulnificus which is a halophilic bacterium, found responsible for causing several human infections.

Biochem/physiol Actions

Bile salts are surface-active steroid compounds and their primary physiological function is to aid in the digestion of lipophilic nutrients in intestinal tracts of vertebrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunsil Choi et al.
Frontiers in microbiology, 11, 597515-597515 (2020-11-27)
The BipA (BPI-inducible protein A) protein is ubiquitously conserved in various bacterial species and belongs to the translational GTPase family. Interestingly, the function of Escherichia coli BipA is not essential for cell growth under normal growth conditions. However, cultivation of
Bacterial degradation of bile salts.
Philipp B
Applied Microbiology and Biotechnology, 89 (4), 903-915 (2011)
Bonggyu Min et al.
Frontiers in bioengineering and biotechnology, 8, 828-828 (2020-08-15)
Stress resistance is an important trait expected of lactic acid bacteria used in food manufacturing. Among the various sources of stress, high temperature is a key factor that interrupts bacterial growth. In this regards, constant efforts are made for the
Luis Noriega et al.
International journal of food microbiology, 94(1), 79-86 (2004-06-03)
Six derivatives with increased resistance to ox gall (MIC: > or = 1% w/v) and one derivative resistant to sodium cholate (MIC: 0.8% w/v) were obtained from more sensitive original Bifidobacterium strains. These microorganisms, and two additional cholate resistant derivatives
D L Scheusner et al.
Applied microbiology, 21(1), 46-49 (1971-01-01)
A population of Escherichia coli ML30 cells was exposed to a quaternary ammonium compound, and injury to the cells was measured by a comparison of counts on Trypticase Soy Agar and Violet Red Bile Agar. Substantial injury could not be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service