Skip to Content
Merck
All Photos(1)

Key Documents

B1145000

Bromhexine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride, Bromohexine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20Br2N2 · HCl
CAS Number:
Molecular Weight:
412.59
Beilstein:
4848376
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bromhexine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(Cc1cc(Br)cc(Br)c1N)C2CCCCC2

InChI

1S/C14H20Br2N2.ClH/c1-18(12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17;/h7-8,12H,2-6,9,17H2,1H3;1H

InChI key

UCDKONUHZNTQPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bromhexine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mladen Stojanov et al.
International journal of pharmaceutics, 422(1-2), 349-355 (2011-10-27)
Bromhexine hydrochloride (bromhexine) is a mucolytic agent with very low aqueous solubility. However, with addition of cyclodextrins (CD) to the formulation, this disadvantage may be limited and therapeutic doses of bromhexine in solution can be achieved. The interaction of bromhexine
Marcello Nicoletti et al.
Natural product research, 24(14), 1325-1329 (2010-08-31)
The utilisation of NMR fingerprinting is proposed as a rapid, available and reliable method to determine the contamination of herbal products. The presence of nimesulide has been reported recently as the contaminant of P.C. 28 Plus, a product based on
Matthew E Spector et al.
Head & neck, 34(12), 1727-1733 (2012-01-17)
Despite better prognosis, there is a group of oropharyngeal squamous cell carcinoma (SCC) human papillomavirus (HPV)+ patients who experience treatment failure and succumb to distant metastasis. Seventy-eight previously untreated patients nested in a concurrent chemoradiation protocol were reviewed to correlate
Saman Azodi-Deilami et al.
Applied biochemistry and biotechnology, 164(2), 133-147 (2010-11-16)
Imprinted polymers are now being increasingly considered for active biomedical uses such as drug delivery. In this work, the use of molecularly imprinted polymers (MIPs) in designing new drug delivery devices was studied. Imprinted polymers were prepared from methacrylic acid
Superior mesenteric artery aneurysm: importance of sonography as the primary imaging technique for detection.
Radu Badea et al.
Journal of ultrasound in medicine : official journal of the American Institute of Ultrasound in Medicine, 29(10), 1503-1506 (2010-09-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service