Skip to Content
Merck
All Photos(1)

Key Documents

A9797

Supelco

Ambroxol hydrochloride

analytical standard

Synonym(s):

2-Amino-3,5-dibromo-N-(trans-4-hydroxycyclohexyl)benzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18Br2N2O · HCl
CAS Number:
Molecular Weight:
414.56
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2

InChI

1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11-;

InChI key

QNVKOSLOVOTXKF-PFWPSKEQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ambroxol hydrochloride is a mucolytic expectorant and a metabolite of bromhexine. It is used in the treatment of respiratory disorders characterized by viscous or excessive mucus.

Application

Ambroxol hydrochloride standard may be used as a reference standard in the determination of ambroxol hydrochloride in pharmaceutical preparations using reversed-phase sequential injection chromatography (RP-SIC) and reversed-phase high performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Metabolite of bromhexine.
A Nav1.8-preferring sodium channel blocker

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jauch, R., et al.
Arzneimittelforschung, 13, 474-474 (1963)
Wolfram Gaida et al.
Neuropharmacology, 49(8), 1220-1227 (2005-09-27)
Neuropathic pain affects many patients, and treatment today is far from being perfect. Nav1.8 Na(+) channels, which are expressed by small fibre sensory neurons, are promising targets for novel analgesics. Na(+) channel blockers used today, however, show only limited selectivity
Giovanni Ciana et al.
Molecular genetics and metabolism reports, 25, 100678-100678 (2020-12-10)
Gaucher disease (GD) is an autosomal recessive lysosomal storage disorder caused by mutations in the acid β-glucosidase encoding gene (GBA1), resulting in the deficient activity of acid β-glucosidase (GCase). To date, there is no approved treatment for the neurological manifestations
Concise Dictionary of Pharmacological Agents: Properties and Synonyms (2012)
Sequential injection chromatographic determination of ambroxol hydrochloride and doxycycline in pharmaceutical preparations.
Satinsky D, et al.
Talanta, 68(2), 214-218 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service