Skip to Content
Merck
All Photos(2)

Key Documents

93832

Supelco

2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol disodium salt dihydrate

≥95.0% (HPLC), for spectrophotometric det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2

Synonym(s):

5-Bromo-PAPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19BrN4Na2O4S · 2H2O
CAS Number:
Molecular Weight:
537.34
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0% (HPLC)

form

powder

quality

for spectrophotometric det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

O.O.[Na+].[Na+].CCCN(CCCS([O-])(=O)=O)c1ccc(\N=N\c2ccc(Br)cn2)c([O-])c1

InChI

1S/C17H21BrN4O4S.2Na.2H2O/c1-2-8-22(9-3-10-27(24,25)26)14-5-6-15(16(23)11-14)20-21-17-7-4-13(18)12-19-17;;;;/h4-7,11-12,23H,2-3,8-10H2,1H3,(H,24,25,26);;;2*1H2/q;2*+1;;/p-2/b21-20+;;;;

InChI key

WQXVWWKVWNUTOV-MKMLRUMESA-L

Application

2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol disodium salt dihydrate may be used as a reagent for the determination of copper and iron by flow injection-catalytic photometric method using serial flow cell.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent for the direct determination of zinc in human serum by flow-injection spectrophotometric analysis[2]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Hayashibe et al.
Analytical Sciences, 10, 795-795 (1994)
R Ostan et al.
Biogerontology, 7(5-6), 437-447 (2006-10-10)
Immunosenescence features, such as thymic involution, alteration of T-cell repertoire, autoimmunity and accumulation of memory/effector T cells, may be the result, at least in part, of a zinc deficiency, which is often observed during ageing. Zinc, as essential trace element
Determination of Cobalt in Natural Water as a 2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulphopropyl)amino]phenol Chelate by On-Line Preconcentration HPLC with Column-Switching Technique
N. Uehara, K. Fukuda, Y. Shijo
Analytical Sciences, 14, 343-343 (1998)
Reversed-phase Ion-pair Partition Liquid Chromatoguraphy of Chelates with 2-(3,5-Dibromo-2-pyridylazo)-5-[N-ethyl-N-(3-sulphopropyl)amino]phenol and Analogues
C. Ohtsuka, H. Wada, T. Ishizuki, G. Nakagawa
Analytica Chimica Acta, 223, 339-339 (1989)
Noriko Hata et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(7), 925-928 (2008-07-11)
The formation of a liquid organic ion associate in an aqueous sample was applied to the concentration and determination of cadmium in environmental water samples. Cadmium was converted into a complex with 2-(5-bromo-2-pyridylazo)-5-(N-propyl-N-sulfopropylamino)phenol (5-Br-PAPS) in a 40-mL sample solution, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service