Skip to Content
Merck
All Photos(1)

Key Documents

51158

Supelco

Bis(1-methyl-2-imidazolyl)ketone

for photometric determination of Fe(II), ≥98.0% (HPLC)

Synonym(s):

BMIK, Bis(1-methyl-2-imidazolyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N4O
CAS Number:
Molecular Weight:
190.20
Beilstein:
914434
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.0% (HPLC)

form

solid

technique(s)

UV/Vis spectroscopy: suitable

solubility

H2O: 50 mg/mL, clear, colorless to slightly yellow

SMILES string

Cn1ccnc1C(=O)c2nccn2C

InChI

1S/C9H10N4O/c1-12-5-3-10-8(12)7(14)9-11-4-6-13(9)2/h3-6H,1-2H3

InChI key

BBDFYGQFWQRHFH-UHFFFAOYSA-N

Application

Highly selective colorimetric indicator for Fe(II) in aqueous samples.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron-Catalyzed Oxidation of Bis(imidazo1-2-yl)methane into Bis(imidazo1-2-yl) Ketone
C. Cuillot, E. Mulliez, P. Leduc, J.-C. Chottardl
Inorganic Chemistry, 29, 579-581 (1990)
Syntheses and structures of mononuclear manganese(II) complexes bearing bis(1-methylimidazol-2-yl)ketone ligands
L. Peters, M.-F. Tepedino, T. Haas, E. Hubner, U. Zenneck, N. Burzlaff
Inorgorganica Chimica Acta, 362, 2678-2685 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service