Skip to Content
Merck
All Photos(2)

Documents

93129

Supelco

Piperacillin sodium salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N5NaO7S
CAS Number:
Molecular Weight:
539.54
Beilstein:
5373920
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)c4ccccc4)C(=O)C1=O

InChI

1S/C23H27N5O7S.Na/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28;/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34);/q;+1/p-1/t13-,14-,15+,20-;/m1./s1

InChI key

WCMIIGXFCMNQDS-IDYPWDAWSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Piperacillin sodium is a semi-synthetic penicillin which belongs to the β-lactam class of antibiotics with a broad spectrum of antibacterial activity against anaerobic pathogens and both gram positive and gram negative bacteria.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piperacillin therapy for serious bacterial infections.
Winston DJ, et al.
The American Journal of Medicine, 69(2), 255-261 (1980)
Piperacillin sodium: antibacterial spectrum, pharmacokinetics, clinical efficacy, and adverse reactions.
Fortner CL, et al.
Pharmacotherapy, 2(6), 287-298 (1982)
T W Felton et al.
Clinical pharmacology and therapeutics, 96(4), 438-448 (2014-06-14)
Pulmonary infections in critically ill patients are common and are associated with high morbidity and mortality. Piperacillin-tazobactam is a frequently used therapy in critically ill patients with pulmonary infection. Antibiotic concentrations in the lung reflect target-site antibiotic concentrations in patients
Erlangga Yusuf et al.
Journal of critical care, 29(6), 1089-1095 (2014-09-03)
The purpose of this study is to review the rationale of prolonged (ie, extended or continuous) infusion of piperacillin/tazobactam (PIP/TAZ) in critically ill patients and to perform a systematic review that compare the effectiveness of prolonged infusion with intermittent bolus
K-A Dietrich et al.
Deutsche medizinische Wochenschrift (1946), 139(28-29), 1459-1461 (2014-07-02)
A 15-year-old boy presented with painful nodules on his palms and discrete pustules on the forearm. Two days earlier he had taken a bath in a new whirlpool. Bacteriological examination of the pustules revealed Pseudomonas aeruginosa. The cervical and axillary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service