Skip to Content
Merck
All Photos(1)

Documents

91316

Supelco

cis-3-Hexen-1-ol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1719712
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.45 (vs air)

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.44 (lit.)
n20/D 1.440

bp

156-157 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC\C=C/CCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h3-4,7H,2,5-6H2,1H3/b4-3-

InChI key

UFLHIIWVXFIJGU-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam F Cooperband et al.
Journal of economic entomology, 112(2), 671-682 (2019-02-13)
The spotted lanternfly, Lycorma delicatula (White), is an invasive phloem feeder recently introduced into North America that attacks a broad range of woody plants. When feeding in large numbers, they can seriously damage or kill a tree. Their preferred host
Shigehiro Namiki et al.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology, 194(5), 501-515 (2008-04-05)
Pheromone-source orientation behavior can be modified by coexisting plant volatiles. Some host plant volatiles enhance the pheromonal responses of olfactory receptor neurons and increase the sensitivity of orientation behavior in the Lepidoptera species. Although many electrophysiological studies have focused on
Andre F Cruz et al.
Phytochemistry, 78, 72-80 (2012-04-24)
Fusarium diseases cause major economic losses in wheat-based crop rotations. Volatile organic compounds (VOC) in wheat and rotation crops, such as chickpea, may negatively impact pathogenic Fusarium. Using the headspace GC-MS method, 16 VOC were found in greenhouse-grown wheat leaves:
Krista L Ryall et al.
Environmental entomology, 41(3), 648-656 (2012-06-27)
Attraction of emerald ash borer, Agrilus planipennis Fairmaire, to a volatile pheromone was demonstrated in three field experiments using baited green sticky traps. A dose-response curve was generated for male A. planipennis to increasing release rates of (3Z)-dodecen-12-olide ((3Z)-lactone) in
Tsviya Olender et al.
Chemical senses, 37(7), 581-584 (2012-06-15)
Considerable evidence supports the idea that odorant recognition depends on specific sequence variations in olfactory receptor (OR) proteins. Much of this emerges from in vitro screens in heterogenous expression systems. However, the ultimate proof should arise from measurements of odorant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service