Skip to Content
Merck
All Photos(1)

Documents

91220

Sigma-Aldrich

Trichloroisocyanuric acid

technical, ≥95% (CHN)

Synonym(s):

1,3,5-Trichloro-2,4,6-triazinetrione, Symclosene, TCICA, Trichlorocyanuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl3N3O3
CAS Number:
Molecular Weight:
232.41
Beilstein:
202022
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥95% (CHN)

form

powder

mp

249-251 °C (lit.)

SMILES string

ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O

InChI

1S/C3Cl3N3O3/c4-7-1(10)8(5)3(12)9(6)2(7)11

InChI key

YRIZYWQGELRKNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Re: Effect of silver oxide/trichloroisocyanuric acid antimicrobial urinary drainage system on catheter-associated bacteriuria.
H B Burke
The Journal of urology, 141(5), 1213-1213 (1989-05-01)
Cristobal Chaidez et al.
International journal of environmental health research, 13(3), 295-302 (2003-08-12)
Outbreaks of pathogenic bacteria infections associated with the consumption of fresh produce has occurred with increased frequency in recent years. This study was undertaken to determine the efficacy of three commonly used disinfectants in packing-houses of Culiacan, Mexico (sodium hypochlorite
Beer Singh et al.
Journal of hazardous materials, 161(2-3), 933-940 (2008-06-03)
High surface area (887.3m(2)/g) silica nanoparticles were synthesized using aerogel route and thereafter, characterized by N(2)-Brunauer-Emmet-Teller (BET), SEM and TEM techniques. The data indicated the formation of nanoparticles of silica in the size range of 24-75 nm with mesoporous characteristics.
T T Martinez et al.
Journal of toxicology. Clinical toxicology, 33(4), 349-354 (1995-01-01)
Two patients were admitted to the hospital emergency room in respiratory distress after an accidental explosion involving chlorinating agents for the swimming pool. The two primary agents involved were calcium hypochlorite and trichloro-s-triazinetrione; both are commonly used chlorinating agents. These
A J Schaeffer et al.
The Journal of urology, 139(1), 69-73 (1988-01-01)
We assessed the efficacy of silver oxide coating of the indwelling urethral catheter and catheter adapter, and instillation of trichloroisocyanuric acid into the urinary drainage bag in the prevention of catheter-associated bacteriuria in a prospective and randomized study of 74

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service