Skip to Content
Merck
All Photos(4)

Key Documents

12060

Sigma-Aldrich

Benzalkonium chloride

≥95.0% ((calculated on dry substance), T)

Synonym(s):

Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
CAS Number:
Beilstein:
4062599
EC Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

description

cationic

Quality Level

Assay

≥95.0% ((calculated on dry substance), T)

form

semisolid

composition

benzyldimethyldodecylammonium chloride, ~70% (Purity (HPLC Area %))
benzyldimethyltetradecylammonium chloride, ~30% (HPLC Minor Component (Area %))

impurities

≤10% water

ign. residue

≤0.1% (as SO4)

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzalkonium chloride is a quaternary ammoniumcompound (QAC) that is most commonly used as a phase transfer catalyst,cationic surfactant, and biocide. It has shown promising applications inchemical synthesis, catalysis, and drug discovery.

Application

Benzalkonium chloride has been used as a:

  • Modifier for montmorillonite clay, enabling the synthesis of eco-friendly and proficient heterogeneous nano-catalysts for green synthesis of 5-substituted 1H-tetrazoles
  • Precursor for the synthesis of alkaline ionic liquids, which have been employed as supported ionic liquid catalysts (SILCAs)
  • Catalyst in the synthesis of functionalized quinoxalines in water

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzalkonium chloride: selective inhibitor of histamine release induced by compound 48/80 and other polyamines.
Read GW & Kiefer EF
Journal of Pharmacology and Experimental Therapeutics, 211(3), 711-715 (1979)
Benzalkonium chloride: selective inhibitor of histamine release induced by compound 48/80 and other polyamines.
Read G W
Journal of Pharmacology and Experimental Therapeutics, 211(3), 711-715 (1979)
Immobilized Cu(0) nanoparticles on montmorillonite-modified with benzalkonium chloride (MMT-BAC@Cu(0)): as an eco-friendly and proficient heterogeneous nano-catalyst for green synthesis of 5-substituted 1H-tetrazoles
Priani F. et al.
Royal Society of Chemistry Advances, 13, 6160-6170 (2023)
Surfactant micelles as microreactors for the synthesis of quinoxalines in water: scope and limitations of surfactant catalysis
Kumar D.
Royal Society of Chemistry Advances, 3, 15157?15168- 15157?15168 (2013)
Accelerated solvent extraction followed by on-line solid-phase extraction coupled to ion trap LC/MS/MS for analysis of benzalkonium chlorides in sediment samples.
Ferrer I & Furlong ET
Analytical Chemistry, 74(6), 1275-1280 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service