90404
Thioglycolate Broth with Resazurine
suitable for microbiology, NutriSelect® Plus
Synonym(s):
Fluid Thioglycolate Medium
About This Item
Recommended Products
Agency
according to ISO 7937:2004
Quality Level
sterility
non-sterile
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 0.75 g/L
casein enzymic hydrolysate, 15.00 g/L
L-cystine, 0.50 g/L
dextrose, 5.50 g/L
resazurine, 0.001 g/L
sodium chloride, 2.50 g/L
sodium thioglycolate, 0.50 g/L
yeast extract, 5.00 g/L
manufacturer/tradename
NutriSelect® Plus
final pH
7.1±0.2
application(s)
agriculture
clinical testing
environmental
food and beverages
sterility testing
microbiology
suitability
Micrococcus spp.
nonselective for Bacillus spp.
nonselective for Bacteroides spp.
nonselective for Candida spp.
nonselective for Clostridium spp.
nonselective for Neisseria spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
Application
Preparation Note
Other Notes
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service