Skip to Content
Merck
All Photos(2)

Key Documents

90107

Sigma-Aldrich

Gram′s iodine solution

for microscopy

Synonym(s):

Potassium Triiodide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

grade

for microscopy

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

color

brown to very dark brown

refractive index

n20/D 1.335

density

1.005 g/mL at 20 °C

suitability

suitable for microscopy
bacteria

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

Gram’s iodine solution is used as a mordant in the Gram staining process. It is added after the primary staining step with Crystal Violet; the resultant Crystal Violet - Iodine complex imparts purple color to the cells.

Biochem/physiol Actions

Iodine(I) interacts with Crystal Violet(CV) to form large CV-I complexes within the cytoplasm and outer layers of the cell.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical Methods for Environmental Microbiology and Biotechnology (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service