Skip to Content
Merck
All Photos(1)

Documents

L6146

Sigma-Aldrich

Lugol solution

Synonym(s):

Iodine/Potassium iodide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.25

General description

Lugol solution is comprised of aqueous potassium iodide plus iodine.

Application

Lugol solution has been used in a study to assess narrow-band imaging without image magnification for detecting high-grade dysplasia and intramucosal esophageal squamous cell carcinoma. It has also been used in a study to determine its benefit in aiding detection of the presence and spread of small squamous cell carcinomas of the esophagus.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo López-Rosales et al.
Toxins, 6(1), 229-253 (2014-01-08)
Benthic marine dioflagellate microalgae belonging to the genus Prorocentrum are a major source of okadaic acid (OA), OA analogues and polyketides. However, dinoflagellates produce these valuable toxins and bioactives in tiny quantities, and they grow slowly compared to other commercially
Jason B Thomas et al.
BMC plant biology, 17(1), 201-201 (2017-11-16)
Pennycress [Thlaspi arvense L (Brassicaceae)] is being domesticated as a renewable biodiesel feedstock that also provides crucial ecosystems services, including as a nutritional resource for pollinators. However, its flowers produce significantly less nectar than other crop relatives in the Brassicaceae.
Stephanie E Lindsey et al.
Development (Cambridge, England), 145(20) (2018-10-20)
Growth and remodeling of the primitive pharyngeal arch artery (PAA) network into the extracardiac great vessels is poorly understood but a major source of clinically serious malformations. Undisrupted blood flow is required for normal PAA development, yet specific relationships between
Edson Ide et al.
World journal of gastroenterology, 17(39), 4408-4413 (2011-11-24)
To compare narrow-band imaging (NBI) without image magnification, and chromoendoscopy with Lugol's solution for detecting high-grade dysplasia and intramucosal esophageal squamous cell carcinoma (SCC) in patients with head and neck cancer. This was a prospective observational study of 129 patients
Michèle G DuVal et al.
PloS one, 9(2), e89183-e89183 (2014-03-04)
Mutation of Glass bottom boat, the Drosophila homologue of the bone morphogenetic protein or growth/differentiation factor (BMP/GDF) family of genes in vertebrates, has been shown to disrupt development of neuromuscular junctions (NMJ). Here we tested whether this same conclusion can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service