Skip to Content
Merck
All Photos(2)

Documents

85640

Millipore

SS-Agar

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Salmonella Shigella Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
brilliant green, 0.0003 g/L
ferric citrate, 1.0 g/L
lactose, 10.0 g/L
meat extract, 5.0 g/L
neutral red, 0.025 g/L
ox bile, dehydrated, 8.5 g/L
peptone, 5.0 g/L
sodium citrate, 10.0 g/L
sodium thiosulfate, 8.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective and differential for Salmonella spp.
selective and differential for Shigella spp.

Application

A differential, selective medium recommended for the isolation of Salmonella and Shigella from stool, foods and clinical material.

Preparation Note

Suspend 63 g in 1 litre of distilled water. Bring to the boil with frequent agitation and allow to simmer gently to dissolve the agar. Do not autoclave. Cool to about 50°C, mix and pour into petri dishes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P. Gunasekaran
Laboratory Manual In Microbiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service