Skip to Content
Merck
All Photos(5)

Documents

H6269

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

for molecular biology, ≥99%

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein:
3598189
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

description

cationic

Assay

≥99%

form

powder or crystals

mol wt

micellar avg mol wt 62,000

aggregation number

170

pH

5.0-7 (25 °C, 36.4 g/L)

CMC

0.92
1 mM (20-25°C)

mp

248-251 °C (lit.)

solubility

H2O: 1/10 g/mL

HLB

10

suitability

suitable for DNA precipitation

foreign activity

DNAse (Detection)
NICKase Detection
RNAse, none detected

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecyltrimethylammonium bromide has been used:

  • as a component of DNA extraction buffer and RNA extraction buffer
  • as a constituent of dye dissolution buffer

Biochem/physiol Actions

Hexadecyltrimethylammonium bromide also called cetyltrimethylammonium bromide (CTAB), is a cationic surfactant. CTAB method is an efficient, cost and time effective method of DNA extraction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

471.2 °F - closed cup

Flash Point(C)

244 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijun Xu et al.
Materials science & engineering. C, Materials for biological applications, 103, 109854-109854 (2019-07-28)
Combination of different therapy modalities with multiple tumoricidal mechanisms has emerged as a promising anticancer strategy. Herein, we reported an aldehyde/catechol-functionalized hyaluronic acid (DAHA) and hydroxyethyl chitosan (HECS) decorated gold nanorod (GNR) platform for combined chemo-photothermal therapy of breast cancer.
Investigation of the Orientation of C T A
Karaca S, et al.
Journal of Chemistry, 2013 (2012)
Xanthoferrin Siderophore Estimation from the Cell-free Culture Supernatants of Different Xanthomonas Strains by HPLC
Pandey SS, et al.
Archives of Virology, 7, e2410-e2410 (2017)
Rolling circle amplification to screen yam germplasm for badnavirus infections and to amplify and characterise novel badnavirus genomes
Bomer M, et al.
Bio-protocol, 8(1), 11-17 (2018)
Modification of a CTAB DNA extraction protocol for plants containing high polysaccharide and polyphenol components
Porebski S, et al.
Plant Molecular Biology Reporter, 15(1), 8-15 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service